The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194272.1 | NP_919248.1 | 2 - 3 | 414 | 110 |
GenBank mRNA | AK127873.1 | - | 2 - 3 | 430 | 110 |
AY369207.1 | - | 2 - 3 | 414 | 110 | |
BC131612.1 | - | 1 - 2 | 66 | 110 | |
BC148208.1 | - | 1 - 2 | 66 | 110 |