The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146319.1 | NP_001139791.1 | 11 - 12 | 1339 | 111 |
NM_001146320.1 | NP_001139792.1 | 10 - 11 | 1376 | 111 | |
NM_001146321.1 | NP_001139793.1 | 12 - 13 | 1531 | 111 | |
NM_001146322.1 | NP_001139794.1 | 11 - 12 | 1150 | 111 | |
NM_023927.2 | NP_076416.2 | 11 - 12 | 1488 | 111 | |
XM_005272057.3 | XP_005272114.1 | 11 - 12 | 1229 | 111 | |
XM_005272058.3 | XP_005272115.1 | 11 - 12 | 1226 | 111 | |
XM_005272059.4 | XP_005272116.1 | 12 - 13 | 1619 | 111 | |
XM_005272060.3 | XP_005272117.1 | 11 - 12 | 1316 | 111 | |
XM_005272061.3 | XP_005272118.1 | 11 - 12 | 1311 | 111 | |
XM_005272062.3 | XP_005272119.1 | 11 - 12 | 1277 | 111 | |
XM_005272063.3 | XP_005272120.1 | 11 - 12 | 1423 | 111 | |
XM_005272064.3 | XP_005272121.1 | 12 - 13 | 1336 | 111 | |
XM_011543591.2 | XP_011541893.1 | 12 - 13 | 2207 | 111 | |
XM_011543592.2 | XP_011541894.1 | 12 - 13 | 2200 | 111 | |
XM_011543593.2 | XP_011541895.1 | 11 - 12 | 1316 | 111 | |
XM_011543594.2 | XP_011541896.1 | 11 - 12 | 1088 | 111 | |
XM_011543595.2 | XP_011541897.1 | 10 - 11 | 1083 | 111 | |
XM_017009781.1 | XP_016865270.1 | 12 - 13 | 1094 | 111 | |
XM_017009782.1 | XP_016865271.1 | 12 - 13 | 1333 | 111 | |
XM_017009783.1 | XP_016865272.1 | 10 - 11 | 944 | 111 | |
XM_017009784.1 | XP_016865273.1 | 10 - 11 | 1135 | 111 | |
XM_017009785.1 | XP_016865274.1 | 10 - 11 | 1185 | 111 | |
GenBank mRNA | AK024966.1 | - | 11 - 12 | 1256 | 111 |
AK225829.1 | - | 11 - 12 | 1256 | 111 | |
AK293307.1 | - | 10 - 11 | 1081 | 111 | |
AK294656.1 | - | 11 - 12 | 1141 | 111 | |
AK296267.1 | - | 12 - 13 | 1236 | 111 | |
AK298009.1 | - | 11 - 12 | 1073 | 111 | |
AK300154.1 | - | 11 - 12 | 1284 | 111 | |
AK303857.1 | - | 11 - 12 | 1090 | 111 | |
AK316288.1 | - | 11 - 12 | 1303 | 111 | |
AY116970.1 | - | 11 - 12 | 1058 | 111 | |
BC008590.1 | - | 11 - 12 | 1221 | 111 |