The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105529.1 | NP_001098999.1 | 1 - 2 | 284 | 77 |
NM_006095.2 | NP_006086.1 | 1 - 2 | 284 | 77 | |
XM_005248043.2 | XP_005248100.1 | 1 - 2 | 279 | 77 | |
XM_011513615.1 | XP_011511917.1 | 1 - 2 | 279 | 77 | |
XM_011513616.2 | XP_011511918.1 | 1 - 2 | 313 | 77 | |
XM_011513618.1 | XP_011511920.1 | 1 - 2 | 279 | 77 | |
XM_017007645.1 | XP_016863134.1 | 1 - 2 | 313 | 77 | |
XM_017007646.1 | XP_016863135.1 | 1 - 2 | 260 | 77 | |
XM_017007647.1 | XP_016863136.1 | 1 - 2 | 258 | 77 | |
GenBank mRNA | AB013452.1 | - | 1 - 2 | 65 | 77 |
AB209687.1 | - | 1 - 2 | 137 | 77 | |
AF067820.1 | - | 1 - 2 | 52 | 77 | |
BC020943.1 | - | 1 - 2 | 272 | 77 | |
BC109317.1 | - | 1 - 2 | 46 | 77 | |
BC109318.1 | - | 1 - 2 | 52 | 77 | |
BP232466.1 | - | 1 - 2 | 284 | 77 |