Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP8A1
Entrez Gene ID
10396
Gene Name
ATPase phospholipid transporting 8A1
Gene Aliases
ATPASEII, ATPIA, ATPP2
Chromosome Location
Chr.4: 42408373 - 42657134 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105529.1 NP_001098999.1 35 - 36 3584 141
NM_006095.2 NP_006086.1 36 - 37 3629 141
XM_005248043.2 XP_005248100.1 36 - 37 3624 141
XM_011513615.1 XP_011511917.1 37 - 38 3645 141
XM_011513616.2 XP_011511918.1 37 - 38 3679 141
XM_011513618.1 XP_011511920.1 36 - 37 3600 141
XM_017007645.1 XP_016863134.1 35 - 36 3613 141
XM_017007646.1 XP_016863135.1 36 - 37 3552 141
XM_017007647.1 XP_016863136.1 34 - 35 3484 141
GenBank mRNA AB013452.1 - 36 - 37 3410 141
AB209687.1 - 35 - 36 3437 141
AF067820.1 - 36 - 37 3397 141
BC109317.1 - 35 - 36 3346 141
BC109318.1 - 35 - 36 3352 141
BX648675.1 - 13 - 14 1277 141

View Full Product Details