The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105529.1 | NP_001098999.1 | 3 - 4 | 502 | 122 |
NM_006095.2 | NP_006086.1 | 3 - 4 | 502 | 122 | |
XM_005248043.2 | XP_005248100.1 | 3 - 4 | 497 | 122 | |
XM_011513615.1 | XP_011511917.1 | 3 - 4 | 497 | 122 | |
XM_011513616.2 | XP_011511918.1 | 3 - 4 | 531 | 122 | |
XM_011513618.1 | XP_011511920.1 | 3 - 4 | 497 | 122 | |
XM_017007645.1 | XP_016863134.1 | 3 - 4 | 531 | 122 | |
XM_017007646.1 | XP_016863135.1 | 3 - 4 | 478 | 122 | |
XM_017007647.1 | XP_016863136.1 | 3 - 4 | 476 | 122 | |
GenBank mRNA | AB013452.1 | - | 3 - 4 | 283 | 122 |
AB209687.1 | - | 3 - 4 | 355 | 122 | |
AF067820.1 | - | 3 - 4 | 270 | 122 | |
BC109317.1 | - | 3 - 4 | 264 | 122 | |
BC109318.1 | - | 3 - 4 | 270 | 122 | |
BP232466.1 | - | 3 - 4 | 502 | 122 |