The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105529.1 | NP_001098999.1 | 8 - 9 | 821 | 96 |
NM_006095.2 | NP_006086.1 | 8 - 9 | 821 | 96 | |
XM_005248043.2 | XP_005248100.1 | 8 - 9 | 816 | 96 | |
XM_011513615.1 | XP_011511917.1 | 8 - 9 | 816 | 96 | |
XM_011513616.2 | XP_011511918.1 | 8 - 9 | 850 | 96 | |
XM_011513618.1 | XP_011511920.1 | 8 - 9 | 816 | 96 | |
XM_017007645.1 | XP_016863134.1 | 8 - 9 | 850 | 96 | |
XM_017007646.1 | XP_016863135.1 | 7 - 8 | 723 | 96 | |
XM_017007647.1 | XP_016863136.1 | 7 - 8 | 721 | 96 | |
GenBank mRNA | AB013452.1 | - | 8 - 9 | 602 | 96 |
AB209687.1 | - | 8 - 9 | 674 | 96 | |
AF067820.1 | - | 8 - 9 | 589 | 96 | |
BC109317.1 | - | 8 - 9 | 583 | 96 | |
BC109318.1 | - | 8 - 9 | 589 | 96 |