Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMYD3
Entrez Gene ID
64754
Gene Name
SET and MYND domain containing 3
Gene Aliases
KMT3E, ZMYND1, ZNFN3A1, bA74P14.1
Chromosome Location
Chr.1: 245749340 - 246507342 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011544260.1 XP_011542562.1 1 - 2 161 140

View Full Product Details