The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011547.2 | NP_001011547.2 | 9 - 10 | 1195 | 141 |
NM_001135181.1 | NP_001128653.1 | 10 - 11 | 1270 | 141 | |
XM_011540924.2 | XP_011539226.1 | 9 - 10 | 2876 | 141 | |
XM_011540925.2 | XP_011539227.1 | 9 - 10 | 2955 | 141 | |
XM_011540926.2 | XP_011539228.1 | 8 - 9 | 1456 | 141 | |
XM_011540927.1 | XP_011539229.1 | 7 - 8 | 1054 | 141 | |
XM_011540928.1 | XP_011539230.1 | 9 - 10 | 1074 | 141 | |
XM_017000558.1 | XP_016856047.1 | 9 - 10 | 2958 | 141 | |
GenBank mRNA | AK131200.1 | - | 9 - 10 | 1148 | 141 |
AK296133.1 | - | 8 - 9 | 868 | 141 | |
BC104857.1 | - | 9 - 10 | 1195 | 141 | |
BC104863.1 | - | 9 - 10 | 1195 | 141 | |
BC143570.1 | - | 10 - 11 | 1258 | 141 | |
BC143572.1 | - | 10 - 11 | 1268 | 141 | |
BC143587.1 | - | 8 - 9 | 989 | 141 | |
BX648549.1 | - | 10 - 11 | 1224 | 141 |