The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261463.1 | NP_001248392.1 | 17 - 18 | 1924 | 150 |
NM_001322945.1 | NP_001309874.1 | 17 - 18 | 1924 | 150 | |
NM_006175.4 | NP_006166.3 | 16 - 17 | 1819 | 150 | |
NM_198060.3 | NP_932326.2 | 17 - 18 | 1924 | 150 | |
XM_005269864.2 | XP_005269921.1 | 16 - 17 | 1795 | 150 | |
XM_005269865.2 | XP_005269922.1 | 17 - 18 | 1900 | 150 | |
XM_005269867.2 | XP_005269924.1 | 17 - 18 | 1900 | 150 | |
XM_006717870.2 | XP_006717933.1 | 17 - 18 | 1900 | 150 | |
XM_011539832.2 | XP_011538134.1 | 17 - 18 | 1900 | 150 | |
GenBank mRNA | AK056605.1 | - | 9 - 10 | 934 | 150 |
AL831980.2 | - | 17 - 18 | 1851 | 150 | |
AL832025.2 | - | 17 - 18 | 1851 | 150 | |
AL832330.2 | - | 17 - 18 | 1851 | 150 | |
AL832457.3 | - | 3 - 4 | 276 | 150 | |
AY081943.2 | - | 16 - 17 | 1796 | 150 | |
AY177620.1 | - | 17 - 18 | 1877 | 150 | |
AY177621.1 | - | 16 - 17 | 1772 | 150 | |
BC126407.1 | - | 17 - 18 | 1760 | 150 | |
BC143751.1 | - | 17 - 18 | 1760 | 150 | |
BX640682.2 | - | 17 - 18 | 1851 | 150 | |
BX640730.1 | - | 17 - 18 | 2008 | 150 | |
BX641052.1 | - | 17 - 18 | 1888 | 150 | |
BX647658.1 | - | 17 - 18 | 1925 | 150 |