The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261463.1 | NP_001248392.1 | 32 - 33 | 3893 | 85 |
NM_001322945.1 | NP_001309874.1 | 31 - 32 | 3785 | 85 | |
NM_006175.4 | NP_006166.3 | 31 - 32 | 3788 | 85 | |
NM_198060.3 | NP_932326.2 | 32 - 33 | 3893 | 85 | |
XM_005269864.2 | XP_005269921.1 | 31 - 32 | 3764 | 85 | |
XM_005269865.2 | XP_005269922.1 | 31 - 32 | 3761 | 85 | |
XM_005269867.2 | XP_005269924.1 | 32 - 33 | 3869 | 85 | |
XM_006717870.2 | XP_006717933.1 | 31 - 32 | 3764 | 85 | |
XM_011539832.2 | XP_011538134.1 | 32 - 33 | 3869 | 85 | |
GenBank mRNA | AL831980.2 | - | 32 - 33 | 3820 | 85 |
AL832330.2 | - | 32 - 33 | 3820 | 85 | |
AL832457.3 | - | 18 - 19 | 2246 | 85 | |
AY081943.2 | - | 31 - 32 | 3765 | 85 | |
AY177620.1 | - | 32 - 33 | 3846 | 85 | |
AY177621.1 | - | 31 - 32 | 3741 | 85 | |
BC126407.1 | - | 32 - 33 | 3729 | 85 | |
BC143751.1 | - | 32 - 33 | 3729 | 85 | |
BX640682.2 | - | 32 - 33 | 3820 | 85 | |
BX640730.1 | - | 32 - 33 | 3977 | 85 | |
BX641052.1 | - | 32 - 33 | 3857 | 85 | |
BX647658.1 | - | 32 - 33 | 3894 | 85 |