Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSTA3
Entrez Gene ID
7264
Gene Name
tissue specific transplantation antigen P35B
Gene Aliases
FX, P35B, SDR4E1
Chromosome Location
Chr.8: 143612618 - 143618043 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317783.1 NP_001304712.1 9 - 10 870 127
NM_003313.3 NP_003304.1 9 - 10 907 127
XM_005251051.3 XP_005251108.2 9 - 10 842 127
XM_011517269.1 XP_011515571.1 9 - 10 934 127
GenBank mRNA AK096752.1 - 7 - 8 1769 127
AK313560.1 - 9 - 10 849 127
BC001941.1 - 9 - 10 843 127
BC093061.1 - 9 - 10 884 127
U58766.1 - 8 - 9 881 127

View Full Product Details