The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172703.2 | NP_001166174.1 | 9 - 10 | 898 | 123 |
NM_001172704.2 | NP_001166175.1 | 8 - 9 | 799 | 123 | |
NM_001291641.1 | NP_001278570.1 | 10 - 11 | 1069 | 123 | |
NM_001291642.1 | NP_001278571.1 | 6 - 7 | 640 | 123 | |
NM_014001.4 | NP_054720.1 | 8 - 9 | 801 | 123 | |
NM_138619.3 | NP_619525.1 | 9 - 10 | 900 | 123 | |
XM_011524563.2 | XP_011522865.1 | 9 - 10 | 874 | 123 | |
XM_017024385.1 | XP_016879874.1 | 9 - 10 | 874 | 123 | |
XM_017024386.1 | XP_016879875.1 | 8 - 9 | 763 | 123 | |
XM_017024387.1 | XP_016879876.1 | 10 - 11 | 1023 | 123 | |
XM_017024388.1 | XP_016879877.1 | 10 - 11 | 1035 | 123 | |
XM_017024389.1 | XP_016879878.1 | 9 - 10 | 868 | 123 | |
XM_017024390.1 | XP_016879879.1 | 8 - 9 | 767 | 123 | |
XM_017024391.1 | XP_016879880.1 | 8 - 9 | 753 | 123 | |
XM_017024392.1 | XP_016879881.1 | 8 - 9 | 822 | 123 | |
GenBank mRNA | AF190864.1 | - | 8 - 9 | 735 | 123 |
AF219138.1 | - | 9 - 10 | 835 | 123 | |
AF219139.1 | - | 8 - 9 | 736 | 123 | |
AK291651.1 | - | 9 - 10 | 865 | 123 | |
AK291685.1 | - | 8 - 9 | 732 | 123 | |
AK293291.1 | - | 8 - 9 | 761 | 123 | |
AK296843.1 | - | 6 - 7 | 573 | 123 | |
AK301895.1 | - | 8 - 9 | 767 | 123 | |
AK302278.1 | - | 9 - 10 | 868 | 123 | |
AL833603.1 | - | 10 - 11 | 1009 | 123 | |
BC063285.1 | - | 8 - 9 | 1501 | 123 | |
BC070044.1 | - | 8 - 9 | 740 | 123 | |
D63876.1 | - | 8 - 9 | 735 | 123 |