The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172703.2 | NP_001166174.1 | 15 - 16 | 2022 | 108 |
NM_001172704.2 | NP_001166175.1 | 14 - 15 | 1923 | 108 | |
NM_001291641.1 | NP_001278570.1 | 16 - 17 | 2193 | 108 | |
NM_001291642.1 | NP_001278571.1 | 12 - 13 | 1764 | 108 | |
NM_014001.4 | NP_054720.1 | 14 - 15 | 1925 | 108 | |
NM_138619.3 | NP_619525.1 | 15 - 16 | 2024 | 108 | |
XM_011524563.2 | XP_011522865.1 | 15 - 16 | 1998 | 108 | |
XM_017024385.1 | XP_016879874.1 | 15 - 16 | 1998 | 108 | |
XM_017024386.1 | XP_016879875.1 | 14 - 15 | 1887 | 108 | |
XM_017024387.1 | XP_016879876.1 | 16 - 17 | 2147 | 108 | |
XM_017024388.1 | XP_016879877.1 | 16 - 17 | 2159 | 108 | |
XM_017024389.1 | XP_016879878.1 | 15 - 16 | 1992 | 108 | |
XM_017024390.1 | XP_016879879.1 | 14 - 15 | 1891 | 108 | |
XM_017024391.1 | XP_016879880.1 | 14 - 15 | 1877 | 108 | |
XM_017024392.1 | XP_016879881.1 | 14 - 15 | 1946 | 108 | |
GenBank mRNA | AF190864.1 | - | 14 - 15 | 1859 | 108 |
AF219138.1 | - | 15 - 16 | 1959 | 108 | |
AF219139.1 | - | 14 - 15 | 1860 | 108 | |
AK124541.1 | - | 1 - 2 | 1537 | 108 | |
AK291651.1 | - | 15 - 16 | 1989 | 108 | |
AK291685.1 | - | 14 - 15 | 1856 | 108 | |
AK301895.1 | - | 14 - 15 | 1891 | 108 | |
AK302278.1 | - | 15 - 16 | 1992 | 108 | |
AL833603.1 | - | 16 - 17 | 2133 | 108 | |
BC063285.1 | - | 12 - 13 | 3257 | 108 | |
BC070044.1 | - | 14 - 15 | 1864 | 108 | |
D63876.1 | - | 14 - 15 | 1859 | 108 |