The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278639.1 | NP_001265568.1 | 1 - 2 | 493 | 169 |
GenBank mRNA | CA489474.1 | - | 1 - 2 | 183 | 169 |
DC397929.1 | - | 1 - 2 | 493 | 169 |