The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320755.1 | NP_001307684.1 | 25 - 26 | 3411 | 140 |
NM_003170.4 | NP_003161.2 | 24 - 25 | 3309 | 140 | |
XM_017024978.1 | XP_016880467.1 | 26 - 27 | 3470 | 140 | |
XM_017024979.1 | XP_016880468.1 | 24 - 25 | 3547 | 140 | |
XM_017024980.1 | XP_016880469.1 | 23 - 24 | 3241 | 140 | |
GenBank mRNA | AL833607.1 | - | 25 - 26 | 3412 | 140 |
BC003696.1 | - | 9 - 10 | 1184 | 140 | |
BC017105.1 | - | 4 - 5 | 452 | 140 | |
BC136522.1 | - | 24 - 25 | 3221 | 140 | |
BC136524.1 | - | 24 - 25 | 3221 | 140 | |
BC150268.1 | - | 24 - 25 | 3223 | 140 | |
D79984.1 | - | 24 - 25 | 3223 | 140 |