The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320755.1 | NP_001307684.1 | 7 - 8 | 902 | 135 |
NM_003170.4 | NP_003161.2 | 6 - 7 | 800 | 135 | |
XM_017024978.1 | XP_016880467.1 | 8 - 9 | 961 | 135 | |
XM_017024979.1 | XP_016880468.1 | 6 - 7 | 1038 | 135 | |
XM_017024980.1 | XP_016880469.1 | 5 - 6 | 732 | 135 | |
GenBank mRNA | AL833607.1 | - | 7 - 8 | 903 | 135 |
BC033074.1 | - | 3 - 4 | 328 | 135 | |
BC136522.1 | - | 6 - 7 | 712 | 135 | |
BC136524.1 | - | 6 - 7 | 712 | 135 | |
BC150268.1 | - | 6 - 7 | 714 | 135 | |
D79984.1 | - | 6 - 7 | 714 | 135 |