Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNG7
Entrez Gene ID
59284
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 7
Gene Aliases
-
Chromosome Location
Chr.19: 53909425 - 53943944 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031896.4 NP_114102.2 3 - 4 520 138
XM_017027093.1 XP_016882582.1 4 - 5 547 138
GenBank mRNA AF288387.1 - 3 - 4 440 138
AF361353.1 - 3 - 4 425 138
AF458897.1 - 3 - 4 425 138
BC069332.1 - 3 - 4 440 138
BC093869.1 - 3 - 4 520 138
BC113503.1 - 3 - 4 520 138

View Full Product Details