The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024907.6 | NP_079183.4 | 2 - 3 | 597 | 106 |
NM_148169.2 | NP_680474.1 | 2 - 3 | 484 | 106 | |
NR_104026.1 | 2 - 3 | 597 | 106 | ||
GenBank mRNA | AF386743.1 | - | 2 - 3 | 524 | 106 |
AK057934.1 | - | 2 - 3 | 484 | 106 | |
BC012385.1 | - | 2 - 3 | 435 | 106 | |
BX338290.2 | - | 2 - 3 | 486 | 106 |