Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBC1D10C
Entrez Gene ID
374403
Gene Name
TBC1 domain family member 10C
Gene Aliases
CARABIN, EPI64C
Chromosome Location
Chr.11: 67403718 - 67410090 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256508.1 NP_001243437.1 3 - 4 291 143
NM_198517.3 NP_940919.1 3 - 4 308 143
NR_046266.1 2 - 3 478 143
XM_006718538.3 XP_006718601.1 3 - 4 567 143
XM_006718539.3 XP_006718602.1 3 - 4 550 143
XM_006718540.3 XP_006718603.1 3 - 4 480 143
XM_006718541.3 XP_006718604.1 3 - 4 575 143
XM_006718542.3 XP_006718605.1 3 - 4 558 143
XM_006718543.2 XP_006718606.1 3 - 4 292 143
XM_006718544.3 XP_006718607.1 3 - 4 486 143
XM_011545001.2 XP_011543303.1 2 - 3 513 143
XM_011545002.2 XP_011543304.1 2 - 3 528 143
GenBank mRNA BC022202.1 - 2 - 3 252 143
BC036873.1 - 2 - 3 263 143
BC062999.1 - 3 - 4 280 143
CR979868.1 - 3 - 4 308 143

View Full Product Details