Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBC1D10C
Entrez Gene ID
374403
Gene Name
TBC1 domain family member 10C
Gene Aliases
CARABIN, EPI64C
Chromosome Location
Chr.11: 67403718 - 67410090 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198517.3 NP_940919.1 7 - 8 711 151
XM_006718538.3 XP_006718601.1 7 - 8 970 151
XM_006718539.3 XP_006718602.1 7 - 8 953 151
XM_006718540.3 XP_006718603.1 7 - 8 883 151
XM_006718541.3 XP_006718604.1 7 - 8 974 151
XM_006718542.3 XP_006718605.1 7 - 8 957 151
XM_006718543.2 XP_006718606.1 7 - 8 691 151
XM_006718544.3 XP_006718607.1 7 - 8 885 151
XM_011545001.2 XP_011543303.1 6 - 7 912 151
XM_011545002.2 XP_011543304.1 6 - 7 931 151
GenBank mRNA BC022202.1 - 6 - 7 655 151
BC036873.1 - 6 - 7 666 151
BC062999.1 - 7 - 8 683 151

View Full Product Details