Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNF44
Entrez Gene ID
22838
Gene Name
ring finger protein 44
Gene Aliases
-
Chromosome Location
Chr.5: 176526699 - 176543456 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014901.4 NP_055716.1 3 - 4 821 176
XM_005265840.2 XP_005265897.1 3 - 4 533 176
XM_005265841.4 XP_005265898.1 3 - 4 778 176
XM_005265842.4 XP_005265899.1 3 - 4 763 176
XM_005265843.2 XP_005265900.1 3 - 4 469 176
XM_005265844.3 XP_005265901.1 3 - 4 574 176
XM_005265845.4 XP_005265902.1 3 - 4 712 176
XM_006714831.2 XP_006714894.1 3 - 4 533 176
XM_006714832.2 XP_006714895.1 3 - 4 533 176
XM_011534466.2 XP_011532768.1 3 - 4 536 176
XM_011534467.2 XP_011532769.1 3 - 4 767 176
GenBank mRNA AB029023.1 - 3 - 4 706 176
AK302385.1 - 3 - 4 574 176
BC039833.1 - 3 - 4 777 176
BC063297.1 - 3 - 4 822 176

View Full Product Details