Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC35E3
Entrez Gene ID
55508
Gene Name
solute carrier family 35 member E3
Gene Aliases
BLOV1
Chromosome Location
Chr.12: 68746148 - 68768677 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018656.2 NP_061126.2 5 - 5 2109 112
XM_017019584.1 XP_016875073.1 4 - 4 2006 112
GenBank mRNA AF148713.1 - 5 - 5 2080 112
BE540822.1 - - 66 112

View Full Product Details