The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253881.1 | NP_001240810.1 | 7 - 7 | 862 | 174 |
NM_004269.3 | NP_004260.2 | 8 - 8 | 970 | 174 | |
XM_017015329.1 | XP_016870818.1 | 9 - 9 | 1039 | 174 | |
GenBank mRNA | AK124232.1 | - | 2 - 2 | 2919 | 174 |
BC002878.1 | - | 7 - 7 | 849 | 174 | |
BC064608.1 | - | 8 - 8 | 938 | 174 |