The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306070.1 | NP_001292999.1 | 8 - 8 | 4448 | 98 |
NM_144778.3 | NP_659002.1 | 9 - 9 | 4536 | 98 | |
NM_207304.2 | NP_997187.1 | 7 - 7 | 4405 | 98 | |
XM_005254018.2 | XP_005254075.1 | 10 - 10 | 4875 | 98 | |
XM_006719912.2 | XP_006719975.1 | 9 - 9 | 4780 | 98 | |
XM_011521045.2 | XP_011519347.1 | 9 - 9 | 4387 | 98 | |
XM_017020308.1 | XP_016875797.1 | 10 - 10 | 4482 | 98 | |
XM_017020309.1 | XP_016875798.1 | 8 - 8 | 4317 | 98 | |
GenBank mRNA | AF339782.1 | - | 1 - 1 | 1475 | 98 |
AF491866.1 | - | 9 - 9 | 4486 | 98 | |
CR749802.1 | - | 8 - 8 | 4364 | 98 |