The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308333.1 | NP_001295262.1 | 1 - 1 | 48 | 123 |
NM_022135.3 | NP_071418.2 | 1 - 1 | 48 | 123 | |
XM_005247698.3 | XP_005247755.1 | 1 - 1 | 48 | 123 | |
GenBank mRNA | AK226022.1 | - | 2 - 2 | 113 | 123 |
AK314819.1 | - | 1 - 1 | 49 | 123 | |
BC044929.1 | - | 1 - 1 | 48 | 123 |