The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172241.2 | NP_758441.2 | 1 - 1 | 3121 | 138 |
GenBank mRNA | AF177229.1 | - | 1 - 1 | 73 | 138 |
BC132871.1 | - | 1 - 1 | 3054 | 138 |