Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FLRT3
Entrez Gene ID
23767
Gene Name
fibronectin leucine rich transmembrane protein 3
Gene Aliases
HH21
Chromosome Location
Chr.20: 14322985 - 14337847 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198391.2 NP_938205.1 2 - 3 461 90
GenBank mRNA AK074909.1 - 1 - 2 136 90
AL529481.3 - 2 - 3 418 90

View Full Product Details