The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145053.1 | NP_001138525.1 | 11 - 12 | 2342 | 106 |
NM_198529.3 | NP_940931.2 | 11 - 12 | 2463 | 106 | |
NR_026738.1 | 11 - 12 | 2463 | 106 | ||
XM_011524759.1 | XP_011523061.1 | 11 - 12 | 2469 | 106 | |
XM_011524763.2 | XP_011523065.1 | 11 - 12 | 2469 | 106 | |
XM_011524765.1 | XP_011523067.1 | 11 - 12 | 2469 | 106 | |
XM_011524766.2 | XP_011523068.1 | 6 - 7 | 1409 | 106 | |
XM_011524768.2 | XP_011523070.1 | 11 - 12 | 2469 | 106 | |
XM_017024591.1 | XP_016880080.1 | 8 - 9 | 1548 | 106 | |
XM_017024592.1 | XP_016880081.1 | 11 - 12 | 2463 | 106 | |
GenBank mRNA | AK128126.1 | - | 8 - 9 | 1689 | 106 |
AK299608.1 | - | 11 - 12 | 2295 | 106 | |
AK302745.1 | - | 10 - 11 | 1772 | 106 | |
AK303060.1 | - | 10 - 11 | 1677 | 106 | |
BC110608.1 | - | 8 - 9 | 1666 | 106 | |
BC110609.1 | - | 8 - 9 | 1666 | 106 | |
BC119807.1 | - | 8 - 9 | 1666 | 106 | |
BC137519.1 | - | 11 - 12 | 2299 | 106 | |
BX648689.1 | - | 11 - 12 | 2401 | 106 |