The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198540.2 | NP_940942.1 | 1 - 2 | 131 | 76 |
GenBank mRNA | AY277592.2 | - | 1 - 2 | 131 | 76 |