Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IGFL1
Entrez Gene ID
374918
Gene Name
IGF like family member 1
Gene Aliases
APRG644, UNQ644
Chromosome Location
Chr.19: 46229752 - 46231243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198541.1 NP_940943.1 1 - 2 46 56
GenBank mRNA AY359013.1 - 1 - 2 46 56
AY672111.1 - 1 - 2 44 56

View Full Product Details