Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HFM1
Entrez Gene ID
164045
Gene Name
HFM1, ATP dependent DNA helicase homolog
Gene Aliases
MER3, POF9, SEC63D1, Si-11, Si-11-6, helicase
Chromosome Location
Chr.1: 91260766 - 91408008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017975.4 NP_001017975.4 30 - 31 3491 145
XM_011540849.1 XP_011539151.1 30 - 31 3574 145
XM_011540850.2 XP_011539152.1 30 - 31 3503 145
XM_011540851.1 XP_011539153.1 30 - 31 3458 145
XM_011540852.2 XP_011539154.1 30 - 31 3440 145
XM_011540855.1 XP_011539157.1 28 - 29 3296 145
XM_011540857.1 XP_011539159.1 28 - 29 3055 145
XM_011540859.2 XP_011539161.1 27 - 28 3024 145
XM_017000490.1 XP_016855979.1 29 - 30 3354 145
XM_017000491.1 XP_016855980.1 29 - 30 3303 145
XM_017000492.1 XP_016855981.1 24 - 25 2538 145
XM_017000493.1 XP_016855982.1 18 - 19 1742 145
GenBank mRNA AB044787.1 - 7 - 8 779 145
AB074274.1 - 2 - 3 242 145
AB204867.1 - 30 - 31 3492 145
AK094079.1 - 13 - 14 1318 145
AK294754.1 - 24 - 25 2508 146
AK302582.1 - 26 - 27 2659 145
BC130525.1 - 15 - 16 1437 145
BC144172.1 - 15 - 16 1437 145
BX649037.1 - 11 - 12 1027 145

View Full Product Details