Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH12
Entrez Gene ID
201625
Gene Name
dynein axonemal heavy chain 12
Gene Aliases
DHC3, DLP12, DNAH12L, DNAH7L, DNAHC12, DNAHC3, DNHD2, HDHC3, HL-19, HL19
Chromosome Location
Chr.3: 57293699 - 57545203 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291661.1 NP_001278590.1 10 - 11 1417 75
NM_198564.3 NP_940966.2 10 - 11 1417 75
XM_011533466.1 XP_011531768.1 10 - 11 1414 75
XM_011533467.1 XP_011531769.1 10 - 11 1381 75
XM_011533468.1 XP_011531770.1 10 - 11 1414 75
XM_011533469.1 XP_011531771.1 10 - 11 1414 75
XM_011533471.2 XP_011531773.1 10 - 11 1414 75
XM_011533474.1 XP_011531776.1 10 - 11 1414 75
XM_017005860.1 XP_016861349.1 10 - 11 1414 75
XM_017005861.1 XP_016861350.1 10 - 11 2276 75
XM_017005863.1 XP_016861352.1 10 - 11 1414 75
GenBank mRNA AK126276.1 - 10 - 11 1414 75

View Full Product Details