Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- AQP12A
- Entrez Gene ID
-
375318
- Gene Name
- aquaporin 12A
- Gene Aliases
-
AQP-12, AQP12, AQPX2
- Chromosome Location
-
Chr.2: 240691845 - 240698483 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_198998.2 |
NP_945349.1 |
3 - 4 |
758 |
113 |
GenBank mRNA |
AB040748.1 |
- |
3 - 4 |
758 |
113 |
- Gene Symbol
- AQP12B
- Entrez Gene ID
-
653437
- Gene Name
- aquaporin 12B
- Gene Aliases
-
INSSA3
- Chromosome Location
-
Chr.2: 240676418 - 240685743 on Build GRCh38
View Full Product Details