Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LHFPL3
Entrez Gene ID
375612
Gene Name
lipoma HMGIC fusion partner-like 3
Gene Aliases
LHFPL4
Chromosome Location
Chr.7: 104328656 - 104908558 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199000.2 NP_945351.1 2 - 3 807 121
GenBank mRNA AK124251.1 - 2 - 3 576 121
AK299759.1 - 2 - 3 686 121
AY260763.1 - 2 - 3 807 121
BC129937.1 - 2 - 3 763 121

View Full Product Details