Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRELP
Entrez Gene ID
5549
Gene Name
proline and arginine rich end leucine rich repeat protein
Gene Aliases
MST161, MSTP161, SLRR2A
Chromosome Location
Chr.1: 203475755 - 203491352 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201348.1 NP_958505.1 1 - 2 170 108
GenBank mRNA BI770477.1 - 1 - 2 107 109
DB498999.1 - 1 - 2 139 108

View Full Product Details