Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDCP2
Entrez Gene ID
200008
Gene Name
CUB domain containing protein 2
Gene Aliases
-
Chromosome Location
Chr.1: 54138995 - 54153006 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201546.3 NP_963840.2 1 - 2 170 130

View Full Product Details