The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_205862.1 | NP_995584.1 | 1 - 2 | 147 | 117 |
GenBank mRNA | BM688740.1 | - | 1 - 2 | 147 | 117 |
BM924331.1 | - | 1 - 2 | 165 | 117 |