The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207186.2 | NP_997069.2 | 1 - 1 | 790 | 104 |
GenBank mRNA | AF209506.1 | - | 1 - 1 | 767 | 104 |
BC137024.1 | - | 1 - 1 | 790 | 104 | |
BC137027.1 | - | 1 - 1 | 790 | 104 |