Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC2A
Entrez Gene ID
387836
Gene Name
C-type lectin domain family 2 member A
Gene Aliases
INPE5792, KACL, PILAR, UNQ5792
Chromosome Location
Chr.12: 9882733 - 9932430 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130711.1 NP_001124183.1 1 - 2 111 70
NM_207375.2 NP_997258.1 1 - 2 111 70
XM_011520654.2 XP_011518956.1 1 - 2 160 70
XM_011520656.2 XP_011518958.1 1 - 2 160 70
XM_011520657.2 XP_011518959.1 1 - 2 159 70
GenBank mRNA AY359126.1 - 1 - 2 77 70
EF127467.1 - 1 - 2 111 70
EU095393.1 - 1 - 2 38 70
EU095394.1 - 1 - 2 38 70

View Full Product Details