The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130711.1 | NP_001124183.1 | 3 - 4 | 353 | 64 |
NM_207375.2 | NP_997258.1 | 3 - 4 | 353 | 64 | |
XM_011520654.2 | XP_011518956.1 | 3 - 4 | 402 | 64 | |
XM_011520656.2 | XP_011518958.1 | 3 - 4 | 402 | 64 | |
XM_011520657.2 | XP_011518959.1 | 3 - 4 | 401 | 64 | |
GenBank mRNA | AY359126.1 | - | 3 - 4 | 319 | 64 |
EF127467.1 | - | 3 - 4 | 353 | 64 | |
EU095393.1 | - | 3 - 4 | 280 | 64 | |
EU095394.1 | - | 3 - 4 | 280 | 64 |