Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C16orf47
Entrez Gene ID
388289
Gene Name
chromosome 16 open reading frame 47
Gene Aliases
-
Chromosome Location
Chr.16: 73125865 - 73812449 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207385.1 NP_997268.1 3 - 4 350 55
XM_017023228.1 XP_016878717.1 8 - 9 2032 55
XM_017023229.1 XP_016878718.1 3 - 4 638 55
GenBank mRNA AK129695.1 - 3 - 4 350 55

View Full Product Details