Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AATK-AS1
Entrez Gene ID
388428
Gene Name
AATK antisense RNA 1
Gene Aliases
-
Chromosome Location
Chr.17: 81165507 - 81183164 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027255.1 3 - 4 647 76
GenBank mRNA AK126811.1 - 3 - 4 647 76

View Full Product Details