The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308209.1 | NP_001295138.1 | 3 - 4 | 408 | 158 |
NM_214710.4 | NP_999875.1 | 3 - 4 | 411 | 158 | |
GenBank mRNA | AY358594.1 | - | 3 - 4 | 448 | 158 |
BC137161.1 | - | 3 - 4 | 413 | 158 | |
BC144505.1 | - | 3 - 4 | 410 | 158 |