Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPRX1
Entrez Gene ID
284355
Gene Name
tetrapeptide repeat homeobox 1
Gene Aliases
TPRX
Chromosome Location
Chr.19: 47801243 - 47803604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198479.2 NP_940881.2 1 - 2 100 169
GenBank mRNA AK097640.1 - 1 - 2 100 169
BC137501.1 - 1 - 2 54 169
BC141863.1 - 1 - 2 54 169
BC144673.1 - 1 - 2 54 169
DQ340180.1 - 3 - 4 320 169

View Full Product Details