Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SET
Entrez Gene ID
6418
Gene Name
SET nuclear proto-oncogene
Gene Aliases
2PP2A, I2PP2A, IGAAD, IPP2A2, PHAPII, TAF-I, TAF-IBETA
Chromosome Location
Chr.9: 128683432 - 128696396 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122821.1 NP_001116293.1 8 - 8 2529 164
NM_001248000.1 NP_001234929.1 8 - 8 2304 164
NM_001248001.1 NP_001234930.1 8 - 8 2228 164
NM_003011.3 NP_003002.2 8 - 8 2602 164
XM_017015013.1 XP_016870502.1 7 - 7 3343 164
XM_017015014.1 XP_016870503.1 8 - 8 3022 164
XM_017015015.1 XP_016870504.1 7 - 7 3017 164
XM_017015016.1 XP_016870505.1 7 - 7 3243 164
GenBank mRNA BQ774097.1 - 1 - 1 164
CA419540.1 - 1 - 1 164
M93651.1 - 8 - 8 2253 164

View Full Product Details