Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NT5C
Entrez Gene ID
30833
Gene Name
5', 3'-nucleotidase, cytosolic
Gene Aliases
DNT, DNT1, HEL74, P5N2, PN-I, PN-II, UMPH2, cdN, dNT-1
Chromosome Location
Chr.17: 75130225 - 75131965 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252377.1 NP_001239306.1 1 - 1 61 113
NM_014595.2 NP_055410.1 1 - 1 61 113
NR_045513.1 1 - 1 61 113
XM_011524700.2 XP_011523002.2 1 - 1 231 113
GenBank mRNA AF154829.2 - 1 - 1 48 113
AK000419.1 - 1 - 1 48 113
BQ431723.1 - 1 - 1 61 113
EU794640.1 - 1 - 1 48 113

View Full Product Details