Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD62
Entrez Gene ID
342850
Gene Name
ankyrin repeat domain 62
Gene Aliases
DKFZp779B1634
Chromosome Location
Chr.18: 12093796 - 12181684 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277333.1 NP_001264262.1 10 - 11 1405 114
XM_017025720.1 XP_016881209.1 10 - 11 1458 114
GenBank mRNA BX649021.1 - 5 - 6 500 114
DW010010.1 - 6 - 7 522 114
DW010011.1 - 3 - 4 114

View Full Product Details