Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD62
Entrez Gene ID
342850
Gene Name
ankyrin repeat domain 62
Gene Aliases
DKFZp779B1634
Chromosome Location
Chr.18: 12093796 - 12181684 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277333.1 NP_001264262.1 11 - 12 1615 152
XM_017025720.1 XP_016881209.1 11 - 12 1668 152
GenBank mRNA DW010011.1 - 2 - 3 152

View Full Product Details