Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR2A25
Entrez Gene ID
392138
Gene Name
olfactory receptor family 2 subfamily A member 25
Gene Aliases
OR2A24P, OR2A25P, OR2A27
Chromosome Location
Chr.7: 144074220 - 144075152 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004488.1 NP_001004488.1 1 - 1 461 76
GenBank mRNA BC136814.1 - 1 - 1 499 76
BC136829.1 - 1 - 1 499 76

View Full Product Details