The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322117.1 | NP_001309046.1 | 17 - 18 | 2963 | 72 |
NM_001322118.1 | NP_001309047.1 | 15 - 16 | 2643 | 72 | |
NM_001322119.1 | NP_001309048.1 | 15 - 16 | 2760 | 72 | |
NM_017990.4 | NP_060460.4 | 17 - 18 | 3012 | 72 | |
XM_005256015.2 | XP_005256072.1 | 16 - 17 | 2662 | 72 | |
XM_005256016.3 | XP_005256073.1 | 16 - 17 | 3025 | 72 | |
XM_006721225.3 | XP_006721288.1 | 14 - 15 | 2422 | 72 | |
XM_017023387.1 | XP_016878876.1 | 16 - 17 | 2374 | 72 | |
XM_017023389.1 | XP_016878878.1 | 16 - 17 | 2148 | 72 | |
GenBank mRNA | AB082521.1 | - | 17 - 18 | 3012 | 72 |
AK093391.1 | - | 4 - 5 | 360 | 72 | |
AK292502.1 | - | 17 - 18 | 2266 | 72 | |
AK302987.1 | - | 15 - 16 | 1897 | 72 | |
AK316384.1 | - | 3 - 4 | 267 | 72 | |
BC150251.1 | - | 17 - 18 | 3033 | 72 | |
BX538153.1 | - | 17 - 18 | 2261 | 72 | |
CR627404.1 | - | 12 - 13 | 1506 | 72 |